Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-synuclein Mouse anti-Human,Mouse, Unconjugated, Clone: Syn506, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $444.00
Specifications
Antigen | alpha-synuclein |
---|---|
Clone | Syn506 |
Applications | ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABN25222MI
|
MilliporeSigma
MABN252225UL |
25 μL |
Each for $177.00
|
|
|||||
MABN25221MI
|
MilliporeSigma
MABN2522100UL |
100 μL |
Each for $444.00
|
|
|||||
Description
alpha-synuclein Monoclonal antibody specifically detects alpha-synuclein in Human,Mouse samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.Specifications
alpha-synuclein | |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Non-A beta component of AD amyloid;Non-A4 component of amyloid precursor;NACP | |
Recombinant human alpha-synuclein oxidized/nitrated in vitro with peroxynitrite. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
14.46 kDa calculated |
Syn506 | |
Monoclonal | |
Purified | |
Neuroscience | |
NP_000336.1 | |
SNCA, NACP, PARK1 | |
IgG1 κ | |
Protein G Purified | |
Clone Syn506 is a mouse monoclonal antibody that detects misfolded alpha-synuclein. It targets an epitope within 20 amino acids from the N-terminal region. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title