Learn More
Genetex IncSupplier Diversity Partner Anti-alpha Synuclein (phospho Ser129) antibody DFS Item
Manufacturer:  Genetex IncSupplier Diversity Partner GTX50222
The Anti-alpha Synuclein (phospho Ser129) antibody from GeneTex is a Rabbit Polyclonal antibody to alpha Synuclein (phospho Ser129). The Anti-alpha Synuclein (phospho Ser129) antibody Recognizes Human, Mouse, Rat. The Anti-alpha Synuclein (phospho Ser129) antibody has been validated for the following Applications: ICC/IF, IHC, WB.
Catalog No. NC1467520