Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Terpineol, Practical, 80-90%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
98-55-5
C10H18O
154.25
WUOACPNHFRMFPN-VIFPVBQESA-N
2-[(1R)-4-methylcyclohex-3-en-1-yl]propan-2-ol
CC1=CC[C@@H](CC1)C(C)(C)O
98-55-5
C10H18O
154.25
WUOACPNHFRMFPN-VIFPVBQESA-N
2-[(1R)-4-methylcyclohex-3-en-1-yl]propan-2-ol
CC1=CC[C@@H](CC1)C(C)(C)O

TE107, 98-55-5, C10H18O

Spectrum™ alpha-Terpineol, Practical , is a naturally occurring monoterpene alcohol often isolated from pine oil. The Alpha isomer is one of three isomers, and is generally the most common constituent. It has a pleasenat lilac odor and for this reason is often used in perfumes, cosmetics and flavoring agents.
Packaging Steel Can
Quantity 20 L
CAS Max % 1
Percent Purity 80 to 90%