Alfa Aesar™ alpha-Toluenesulfonyl fluoride, 99%

Manufacturer: Alfa Aesar™ B2214603

Quantity 1g

 View more versions of this product

Catalog No. AAB2214603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 329-98-6
    Molecular Formula C7H7FO2S
    Formula Weight 174.19
    MDL Number MFCD00007424
    UN Number UN2923
    Beilstein 2088311
    Synonym Phenylmethanesulfonyl fluoride; Phenylmethylsulfonyl fluoride
    Chemical Name or Material alpha-Toluenesulfonyl fluoride

    Specifications

    Formula Weight 174.19
    TSCA TSCA
    Molecular Formula C7H7FO2S
    MDL Number MFCD00007424
    UN Number UN2923
    CAS 329-98-6
    Beilstein 2088311
    Synonym Phenylmethanesulfonyl fluoride; Phenylmethylsulfonyl fluoride
    Health Hazard 1 Danger
    Melting Point 92° to 94°C
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    RTECS Number XT8040000
    Chemical Name or Material alpha-Toluenesulfonyl fluoride
    Assay 99%

    Description

    Applications
    α-Toluenesulfonyl fluoride (PMSF) is used in the preparation of cell lysates to inhibit protein digestion. It is a widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations when testing CB1 receptor activity.

    Solubility
    Soluble in ether, benzene, anhydrous ethanol, methanol, 2- propane. Hydrolyses in water.

    Notes
    Moisture sensitive. Store away from strong oxidizing agents, amines, bases, water/ moisture. Store at 4°C

    GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit