Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Tubulin Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$516.35

Specifications

Antigen alpha Tubulin
Host Species Rabbit
Isotype IgG
Gene Alias Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub2, LIS3, TUBA3hum-a-tub1, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific
Target Species Human, Mouse, Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP254687 View Documents Novus Biologicals
NBP254687
100 ul Each for $516.35
Add to cart
 
NBP254708 View Documents Novus Biologicals
NBP254708
100 ul Each for $516.35
Add to cart
 
Description

Description

alpha Tubulin Polyclonal antibody specifically detects alpha Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

alpha Tubulin
IgG
Human, Mouse, Rat
RUO
Affinity Purified
Primary
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Rabbit
Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub2, LIS3, TUBA3hum-a-tub1, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit