Learn More
AlphaA Crystallin/CRYAA Antibody (OTI3B12), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270450UV
Catalog No. NBP270450UV
Add to Cart
Description
Description
AlphaA Crystallin/CRYAA Monoclonal specifically detects AlphaA Crystallin/CRYAA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
AlphaA Crystallin/CRYAA
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
CRYAA
Full length recombinant protein of human AlphaA Crystallin/CRYAA (NP_000385) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG2b
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI3B12
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
CRYA1crystallin, alpha-1, crystallin, alpha A, Heat shock protein beta-4, HSPB4alpha-crystallin A chain, human alphaA-crystallin (CRYA1), 10HspB4
Mouse
Affinity Purified
Membrane Trafficking and Chaperones, Vision
1409.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions