Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaB Crystallin/CRYAB Mouse anti-Bovine, Human, Rat, Clone: 3A10.H4, Novus Biologicals™

Mouse MonoclonalAntibody

$235.00 - $358.00

Specifications

Antigen AlphaB Crystallin/CRYAB
Clone 3A10.H4
Dilution Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
NB59687200U View Documents Novus BiologicalsSupplier Diversity Partner
NBP259687200UG
200 ug Unconjugated Each for $358.00
Add to cart
 
NB5968750UG View Documents Novus BiologicalsSupplier Diversity Partner
NBP25968750UG
50 ug Unconjugated Each for $235.00
Add to cart
 
Description

Description

AlphaB Crystallin/CRYAB Monoclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human, Rat, Bovine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

AlphaB Crystallin/CRYAB
Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:100
Purified
Human, Rat, Bovine
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
Native Alpha B Crystallin
Monoclonal
Protein G purified
Does not cross-react with alphaA-crystallin, BetaL-crystallin, BetaH-crystallin, gamma -crystallin, HSP25, HSP27 or HSP47 proteins.
3A10.H4
Western Blot, Immunocytochemistry, Immunofluorescence
Mouse
RUO
1410
IgG1
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit