Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaB Crystallin/CRYAB Mouse anti-Human, Rat, Allophycocyanin/Cy7, Clone: OTI6A9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270454APCCY7

Catalog No. N270454APC7


Add to cart

Description

Description

AlphaB Crystallin/CRYAB Monoclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

AlphaB Crystallin/CRYAB
OTI6A9
Flow Cytometry
Affinity Purified, Conjugated
CRYAB
Full length recombinant protein of human AlphaB Crystallin/CRYAB (NP_001876) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision
Primary
1410.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit