Learn More
AlphaProTech Critical Cover™ AlphaGuard™ Coveralls i
Fluid repellent and static dissipative
Manufacturer:  AlphaProTech CV740323
 View more versions of this product
Catalog No. 19-130-1502C
  Check Availability
Add to cart
Alert:
Not recommended for use with chemicals.
Specifications Description & Specifications
Specifications
Elastic wrist, ankle and back
25 in.
Elastic
Elastic
White
Suitable for use in controlled environments with low particle count
34 in.
29.5 in.
AlphaGuard™ Fabric
63.5cm
Elastic
Serged
Large
Yes
86.36cm
74.93cm