Learn More
AlphaProTech Critical Cover™ AlphaGuard™ Coveralls i
Fluid repellent and static dissipative
Manufacturer:  AlphaProTech CV740324
 View more versions of this product
Catalog No. 19-130-1502D
  Check Availability
Add to cart
Alert:
Not recommended for use with chemicals.
Specifications Description & Specifications
Specifications
AlphaGuard™ Fabric
Serged
X-Large
68.58cm
Yes
92.71cm
76.2cm
Elastic wrist, ankle and back
White
27 in.
Suitable for use in controlled environments with low particle count
36.5 in.
30 in.