Learn More
AlphaProTech Critical Cover™ AlphaGuard™ Coveralls i
Fluid repellent and static dissipative
Manufacturer:  AlphaProTech CV740326
 View more versions of this product
Catalog No. 19-130-1502F
  Check Availability
Add to cart
Alert:
Not recommended for use with chemicals.
Specifications Description & Specifications
Specifications
Elastic wrist, ankle and back
32 in.
Elastic
Elastic
White
Suitable for use in controlled environments with low particle count
38 in.
32 in.
AlphaGuard™ Fabric
81.28cm
Elastic
Serged
3X-Large
Yes
96.52cm
81.28cm