Learn More
AlphaProTech Critical Cover™ AlphaGuard™ Coveralls i
Fluid repellent and static dissipative
Manufacturer:  AlphaProTech CV840327
 View more versions of this product
Catalog No. 19-130-1503G
  Check Availability
Add to cart
Alert:
Not recommended for use with chemicals.
Specifications Description & Specifications
Specifications
AlphaGuard™ Fabric
Serged
4X-Large
82.55cm
Yes
104.14cm
82.55cm
Elastic wrist, ankle and back
Blue
32.5 in.
Suitable for use in controlled environments with low particle count
41 in.
32.5 in.