AlphaProTech Critical Cover™ BarrierTech™ Gowns

Provides protection for the wearer from particles, fluids and light chemical splash

Manufacturer: AlphaProTech GN-22524-2

 View all options for this product

Catalog No. 19-169-4037


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    For Use With (Application) Protection for the wearer from particles, fluids and light chemical splash
    Cuff Style Elastic
    Color White
    Material BarrierTech™ Fabric
    Size Medium

    • Offering a durable and fluid impervious solution for both your lab and/or production areas
    • Low particle count for use in controlled environments
    • Banded ties are attached to the neck and waist for easy and secure closure
    • Serged seams and elastic wrists
    • Available in two colors

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit