AlphaProTech Critical Cover™ ChemTech™ Coveralls Available on GSA/VA Contract for Federal Government customers only.

Designed with a safety barrier on the inside for an added measure of protection

Manufacturer: AlphaProTech CVBK42127

 View more versions of this product

Catalog No. 19-169-5037

  Check Availability
Add to Cart

    Specifications

    Includes Boots, elastic wrist, protective flap over zipper closure
    Seam Type Bound
    Color Yellow
    Size 4X-Large
    For Use With (Application) Chemical barrier, liquid and particle holdout
    Length (English) Sleeve 41 in.
    Length (Metric) Sleeve 104.14cm
    Width (English) Chest 32.5 in.
    Cuff Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit