Learn More
AlphaProTech Critical Cover™ Combo™ Masks with Shield
Manufacturer:  AlphaProTech 5194T
 View more versions of this product
Catalog No. 19-169-4067
Add to cart
Description
Description
  • Fluid resistant mask
  • Shield attaches to mask with snaps
  • 7 mil optically clear shield with anti-fog shield coating, low glare and is distortion free
Specifications
Specifications
Blue
Fluid
Anti-fog coating optically clear full wrap around shield (7 mil thickness) with Foam anti-splash foreheadband, ties