AlphaProTech Critical Cover™ ComforTech™ Coveralls

Lightweight, durable material delivers excellent protection against particles, biologicals and fluids

Manufacturer: AlphaProTech CV-J4192-4

 View all options for this product

Catalog No. 19-125-093


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Closure Type Zipper
    Material ComforTech™ Fabric
    Includes Elastic hood, wrist and ankle
    Seam Type Serged
    Color White
    Size X-Large
    Size (English) Chest 27 in.
    Size (Metric) Chest 68.58cm
    For Use With (Application) Suitable for use in controlled environments and barrier to particles, biologicals and fluids with 99.9% filtration efficiency
    Latex Free Yes
    Length (English) Sleeve 36.5 in.
    Length (Metric) Sleeve 92.71cm
    Size (English) Inseam 30 in.
    Size (Metric) Inseam 76.2cm

    Material rapidly dissipates vapor and heat on a microscopic level which helps to keep wearer cool and comfortable throughout the workday.

    • Material has static dissipative properties
    • Exhibits an ultralow particle count and 99.9% filtration efficiency
    • Fluid and particulate barrier
    • Serged seams
    • Latex-free
    • White

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit