AlphaProTech Critical Cover™ ComforTech™ Frocks

Exceptionally clean with an ultralow particle count

Manufacturer: AlphaProTech FKJ21215

 View more versions of this product

Catalog No. 19-130-4235

  Check Availability
Add to Cart

    Specifications

    Length (Metric) Sleeve 95.25cm
    Size (English) Chest 26.5 in.
    Size (Metric) Chest 67.3cm
    Closure Type Snap
    Color White
    Material ComforTech™ Fabric
    Size 2X-Large

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit