AlphaProTech Critical Cover™ ComforTech™ Sleeves for Sterile Suits

Ultra-clean material to ensure a low particulate count and protection for sterile work areas

Manufacturer: AlphaProTech MS01J764ST

 View more versions of this product

Catalog No. 19-168-457

  Check Availability
Add to Cart

    Specifications

    Cuff Style Elastic
    Length (English) 21.5 in.
    Length (Metric) 54.6cm
    Material Fabric
    Size X-Large
    For Use With (Equipment) Sterile suit
    Sterile Yes
    Color White
    Construction Sonically welded seams and tunneled elastic at both ends

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit