AlphaProTech Critical Cover™ CoolOne™ Mask with Magic Arch™ Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: AlphaProTech YL 6055
 View more versions of this product
Catalog No. 19-169-4065
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications/Compliance ASTM F2299
    Filter Efficiency 95.5%
    Color Yellow
    Pore Size 0.1μ
    Includes Magic Arch™ support strip, soft knit earloop
  • Description & Specifications
Related Products
Product Content Feedback