Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
AlphaProTech Critical Cover™ Coverall Face Shields
BIOPHARMA
Catalog Number 23309828
Click to view available options
Headband Material:
Adjustable Hook-and-Loop-Closure
Adjustable Hook-and-Loop-Closure Headband
Elastic Comfort Band
Model:
Full-face
Half-face
This item is not returnable. View return policy
This item is not returnable. View return policy
Good peripheral vision while acting as a shield against dangerous splashes in the work environment
  • Lightweight and easily fits over glasses
  • Double-sided anti-fog coating
  • Low distorting and 100% optically clear
  • Soft polyurethane foam headband fits snugly against the forehead
  • Full-face style (Cat. Nos. 23-309-829 and 23-309-828) provides complete coverage including the sides of the face
Thickness (English) 7 mil shield
Description Fits easily over glasses
Lens Tint Clear
Headband Material Elastic Comfort Band
No. per Case 100/Cs.
Model Full-face
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Cancel Submit