Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AlphaProTech Critical Cover™ GenPro™ Bouffants
Click to view available options
Size:
21 in.
24 in.
26 in.
Description
- Basic protection for non-hazardous environments
- Non-binding, latex-free elastic
- Secure fit
- Serged seams
- Lightweight material
- White
Specifications
Size | 21 in. |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Model | BO-138 |
Seam Type | Serged |
Quantity | 1000/Cs. |
Includes | Non-binding, Latex-free Band |
For Use With (Application) | Dry, Non-hazardous Environments |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction