AlphaProTech Critical Cover™ GenPro™ Coveralls

Limited barrier to particulates and contamination

Manufacturer: AlphaProTech CV140322

 View more versions of this product

Catalog No. 19-169-4005

  Check Availability
Add to Cart

    Specifications

    Material Microbreathe™ Fabric
    Includes Elastic wrist, ankle and back
    Seam Type Serged
    Color White
    Size Medium
    Size (English) Chest 24.25 in.
    Size (Metric) Chest 61.59cm
    For Use With (Application) Workers and visitors in non-hazardous environments with limited barrier to particulates and contamination
    Latex Free Yes
    Length (English) Sleeve 34 in.
    Length (Metric) Sleeve 86.36cm
    Size (English) Inseam 28.5 in.
    Size (Metric) Inseam 72.39cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit