AlphaProTech Critical Cover™ GenPro™ Lab Coats

Designed for non-hazardous environments

Manufacturer: AlphaProTech LC124219

 View more versions of this product

Catalog No. 19-130-4244

  Check Availability
Add to Cart

    Specifications

    Color White
    Cuff Style Elastic
    Size 4X-Large
    Collar Type Tapered
    Latex Free Yes
    Length (Metric) Sleeve 99.6cm
    Seam Type Serged
    Size (English) Chest 31.5 in.
    Size (Metric) Chest 80cm
    Closure Type Snap
    Length (English) 46.5 in.
    Length (Metric) 118.11cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit