AlphaProTech Critical Cover™ GenPro™ Shoe Covers: Heat-Sealed Seams Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: AlphaProTech SB41222B

 View more versions of this product

Catalog No. 19-169-4140

  Check Availability
Add to Cart

    Specifications

    Size X-Large
    Sole Type Non-skid adhesive
    Product Type Non-conductive Boot Covers
    Material GenPro™ Fabric
    For Use With (Application) Ideal for non-hazardous environments
    Seam Type Heat sealed
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit