AlphaProTech Critical Cover™ GenPro™ Shoe Covers: Serged Seam

For general use environments

Manufacturer: AlphaProTech SH61222B

 View more versions of this product

Catalog No. 19-130-4270

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Size Universal
    Sole Type Non-skid adhesive, serged
    Product Type Non-conductive Boot Covers
    Material GenPro™ Fabric
    For Use With (Application) Ideal for non-hazardous environments
    Height (English) 7 in.
    Height (Metric) 17.78cm
    Length (English) 15.75 in.
    Latex Free Yes
    Length (Metric) 40cm
    Seam Type Serged
    Color Blue
    Thread Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit