AlphaProTech Critical Cover™ MVT Highly Breathable Masks
Provides limited flitration efficiency.
Manufacturer: AlphaProTech 9240
 View more versions of this product
Catalog No. 19-125-370
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance ASTM F2299
    Filter Efficiency 27%
    Color White
    Resistance Fluid
    Pore Size 0.1μ
    Includes Polyethylene tie strings
  • Description & Specifications
Related Products
Product Content Feedback