AlphaProTech Critical Cover™ SafeStep™ Shoe Covers
High-performing, environmentally friendly shoe cover
Manufacturer: AlphaProTech SHE1715BH
 View more versions of this product
Catalog No. 19-125-442
  Check Availability
Add to Cart
    Specifications
    Size Medium
    Product Type SafeStep; Non-skid physical properties
    Material 100% Thermoplastic Polymers; No PVC compounds
    Seam Type Serged
    Thread Color Yellow
    Color White
  • Description & Specifications
Related Products
Product Content Feedback