Learn More
AlphaProTech Critical Cover™ ShieldMate™ Mask with Eye Shield Available on GSA/VA Contract for Federal Government customers only.
Provides eye and face splash protection and inhalation security
Manufacturer:  AlphaProTech BL 8101 S
Catalog No. 19-169-4068
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Features a low glare, anti-fog treated, optically clear shield ultrasonically welded to the mask
  • Magic Arch™ support strip holds the mask away from the nose and mouth to create a larger breathing chamber without limiting downward vision
  • Mask is cooler, more comfortable performance for the wearer
  • Comfortable ties and a full-length aluminum nosepiece comfortably secure the ShieldMate mask to the face
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ASTM F2299
99%
Ultrasonic welded eye shield (5 mil thickness) with anti-fog treatment, comfortable ties, anti-fog Foam strip at the nosepiece, Magic Arch™ support strip
Blue
0.1μ