Learn More
Ansell™ AlphaTec™ 04-414 PVC-Nitrile Blend Coated Gauntlet Gloves
Outstanding chemical protection, comfort PVC glove with superb grip performance
$164.00
Specifications
Color Green
Material Nitrile, PVC
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-014906
View Documents
Ansell™
04414 9 211229
Large Dozen of 12 for $164.00
Case of 6 DZ for $865.50
Add to cart
19-014907
View Documents
Ansell™
04414 10 211230
X-Large Dozen of 12 for $164.00
Case of 6 DZ for $865.50
Add to cart
Description
Description
Ideal for applications involving petrochemicals and for handling equipment and abrasive materials coated with acids, caustics, solvents, grease, and oil.
  • Feature PVC-nitrile blend rough finish
  • Curved, preflexed fingers and wing thumb give extra comfort and flexibility
  • Two-piece lining eliminates seams from work areas to prevent irritation and premature wear-through
Specifications
Specifications
Green
Nitrile, PVC