Learn More
Ansell™ AlphaTec™ 37-310 Latex-Free Food Approved Glove
Ansell™ AlphaTec™ 37-310 Latex-Free Food Approved Glove provides all day comfort at an exceptional value.
$12.05
Specifications
Certifications/Compliance REACH Compliant
Certifications/Compliance (Europe) EN420:2003 + A1:2009; Category lll
Color Blue
Glove Cuff Style Gauntlet
Material Nitrile
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-500-899 View Documents Ansell™
37310070
7 Pack of 12 for $12.05
Case of 12 PK for $115.50
Add to cart
19-500-900 View Documents Ansell™
37310080
8 Pack of 12 for $12.05
Case of 12 PK for $115.50
Add to cart
19-500-901 View Documents Ansell™
37310090
9 Pack of 12 for $12.05
Case of 12 PK for $115.50
Add to cart
19-500-902 View Documents Ansell™
37310100
10 Pack of 12 for $12.05
Case of 12 PK for $115.50
Add to cart
19-500-903 View Documents Ansell™
37310110
11 Pack of 12 for $12.05
Case of 12 PK for $115.50
Add to cart
Description
Description
  • 100% latex free Blue color
  • Unflocked to prevent the risk of food contamination
  • Food Approved
Specifications
Specifications
REACH Compliant
Blue
Nitrile
12 pairs/bag; 12 bags/carton
8 mil
EN420:2003 + A1:2009; Category lll
Gauntlet
AlphaTec™ 37-310
AlphaTec™ 37-310
0.2 mm