Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell AlphaTec™ 87-089 Yellow Lightweight Latex Gloves
Click to view available options
Size:
6.5
7.5
8.5
9.5
Description
- Natural rubber latex
- Abrasion/tear resistant
- 100% cotton flock lining
- Beaded cuff for easy donning
- Ideal for agriculture, food processing or chemical applications that are low hazard risk
Specifications
Certifications/Compliance | FDA 21 CFR 177.2600 |
Certifications/Compliance (Europe) | CE0493, EN420:2003 + A1:2009 |
Chlorinated | Yes |
AQL | 0.65 |
Color | Yellow |
Latex Free | No |
Material | Latex |
Size | 7.5 |
Thickness (English) | 17 mil |
Thickness (Metric) | 0.43 mm |
Show More |