Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-1800 White Coveralls with Collar

Provide the wearer with entry level, low hazard liquid and particulate protection. Ansell™ Alphatec™ MICROCHEM™ 68-1800 White Coveralls with Collar are ideal for a wide range of general industrial applications including general maintenance, paint spraying, powder handling, and food work.

Manufacturer:  Ansell™ WH18B9210304

 View more versions of this product

Catalog No. 17-999-966


  Check Availability
Add to cart

Alert:

This product contains natural rubber latex which may cause allergic reactions.


Specifications

Description

  • Protection:  Low hazard liquid spray, splash and particle protection
  • Comfort:  Moisture vapor permeable ("breathable") to help reduce the risk of heat stress
  • Low Linting:  Reduces the risk of fiber contamination in some critical areas
  • Anti-static:  Tested according to EN1149-5 & AATCC76 (8EMSL test method)
  • Design:  Elastic waist, wrists and ankles, 2-way front zipper with storm flap, bound seams, collar
  • Applications:  General maintenance, paint spraying, powder handling, food
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 8

Description & Specifications

Specifications

2-way front zipper with taped storm-flap
Elasticated wrists
Bound seams
Large
Collar
Particulate and light liquid splash protection
Elasticated
Microporous laminate
White
No
Elasticated
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit