Ansell™ Alphatec™ MICROCHEM™ 68-1800 White Coveralls with Hood and Boots

Provide the wearer with entry level, low hazard liquid and particulate protection. Ansell™ Alphatec™ MICROCHEM™ 68-1800 White Coveralls with Hood and Boots are ideal for a wide range of general industrial applications including general maintenance, paint spraying, powder handling, and food work.

Manufacturer: Ansell™ WH18B9210707

 View more versions of this product

Catalog No. 17-999-977

  Check Availability
Add to Cart

Alert

This product contains natural rubber latex which may cause allergic reactions.


    Specifications

    Material Microporous laminate
    Seam Type Bound seams
    Color White
    Hood Type 3 piece respirator fit hood
    Latex Free No
    Neck Style Attached hood
    Closure Type 2-way front zipper with taped storm-flap
    Waist Style Elasticated
    Protection Class Particulate and light liquid splash protection
    Cuff Style Elasticated wrists
    Size 3XL
    Ankle Style Elasticated with attached anti-skid boots (SureSole step)

    • Protection:  Low hazard liquid spray, splash and particle protection
    • Comfort:  Moisture vapor permeable ("breathable") to help reduce the risk of heat stress
    • Low Linting:  Reduces the risk of fiber contamination in some critical areas
    • Anti-static:  Tested according to EN1149-5 & AATCC76 (8EMSL test method)
    • Design:  Elastic waist, wrists and ankles, 2-way front zipper with storm flap, bound seams, 3 piece respirator fit hood, attached anti-skid boots with SureStep soles
    • Applications:  General maintenance, paint spraying, powder handling, food
    • Sizes:  Available in Small through 5X-Large
    • Certifications:  EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
    • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

    Certifications

    EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 11

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit