Ansell™ Alphatec™ MICROCHEM™ 68-2000 White Coveralls with Hood

Provide breathable protection and comfort with exceptional liquid and particulate protection. Ansell™ Alphatec™ MICROCHEM™ 68-2000 White Coveralls with Hood are ideal for a wide range of industrial applications, with the added benefits of attached 3-piece respirator fit hoods.

Manufacturer: Ansell™ WH20B9211103

 View more versions of this product

Catalog No. 17-999-877

  Check Availability
Add to Cart

Alert

This product contains natural rubber latex which may cause allergic reactions.


    Specifications

    Size Medium
    Hood Type 3 piece respirator fit hood
    Neck Style Attached hood
    Closure Type 2-way front zipper with taped storm-flap
    Protection Class Particulate and light liquid splash protection
    Material Microporous laminate
    Seam Type Bound seams
    Color White
    Latex Free No
    Waist Style Elasticated
    Ankle Style Elasticated
    Cuff Style Elasticated wrists

    • Protection:  Excellent liquid penetration resistance & barrier to fine particulates (0.01µm) including ASTMF1761 and EN14126:2003 resistance to penetration of blood, body fluids and blood-borne pathogens
    • Comfort:  Moisture vapor permeable ("breathable") to help reduce the risk of heat stress
    • Low Linting:  Reduces the risk of fiber contamination in some critical areas
    • Anti-static:  Tested according to EN1149-5 & AATCC76 (8EMSL test method)
    • Design:  Elastic wrists and ankles, 2-way front zipper with storm flap, bound seams, finger loops, 3 piece respirator fit hood
    • Applications:  Pharmaceutical industries, agriculture, cleanrooms, paint spraying, crime scene investigations, veterinary services, food
    • Sizes:  Available in Small through 5X-Large
    • Certifications:  EN 14126 / ASTM F1671, DIN 32781, EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B
    • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

    Certifications

    EN 14126 / ASTM F1671, DIN 32781, EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit