Ansell™ Alphatec™ MICROCHEM™ 68-2300 Yellow Coveralls with Hood

Provide an excellent barrier to harmful chemicals and blood borne pathogens. Ansell™ Alphatec™ MICROCHEM™ 68-2300 Yellow Coveralls with Hood are lighweight and relatively strong and durable. These coveralls are designed for applications such as environmental clean-up, sewage purification installations, and composites manufacturing.

Manufacturer: Ansell™ YY23B9211105

 View more versions of this product

Catalog No. 17-999-911

  Check Availability
Add to Cart

Alert

This product contains natural rubber latex which may cause allergic reactions.


    Specifications

    Material Barrier laminate fabric
    Seam Type Bound seams
    Color Yellow
    Size XL
    Hood Type 3 piece respirator fit hood
    Latex Free No
    Neck Style Attached hood
    Closure Type 2-way front zipper with taped storm-flap
    Waist Style Elasticated
    Protection Class Particulate and light liquid splash protection
    Ankle Style Elasticated
    Cuff Style Elasticated wrists, finger loops

    • Protection:  A barrier to numerous inorganic liquid chemicals including acids and bases; achieves the highest classifications for protection from biological agents in accordance with EN 14126:2003 & ASTM F1671 for penetration of blood, bodily fluids, and blood-borne pathogens
    • Comfort:  Lightweight yet strong and durable
    • Anti-static:  Tested according to EN1149-5 & AATCC76 (8EMSL test method)
    • Design:  Elastic waist, wrists and ankles, 2-way front zipper with taped storm flap, bound seams, finger loops, 3 piece respirator fit hood
    • Applications:  Environmental cleanup, sewage purification installations, composites manufacturing
    • Sizes:  Available in Small through 5X-Large
    • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671
    • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

    Certifications

    EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1674

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit