Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-3000 Yellow Coveralls with Hood

Provide an extremely effective barrier against both chemical and biological hazards. Ansell™ Alphatec™ MICROCHEM™ 68-3000 Yellow Coveralls with Hood are durable and multi-layered, and are some of the most comfortable chemical protective materials available.

$365.50 - $460.50

Specifications

Latex Free No
Color Yellow
Material Barrier laminate fabric
Neck Style Hooded
Hood Type 3-panel, Respirator-fit
View More Specs

Alert:

This product contains natural rubber latex which may cause allergic reactions.


Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-999-924
View Documents Promotion Details
Ansell™
YE30W9211102
Small
Case of 6 for $365.50
Add to cart
 
17-999-925
View Documents Promotion Details
Ansell™
YE30W9211103
Medium
Case of 6 for $365.50
Add to cart
 
17-999-926
View Documents Promotion Details
Ansell™
YE30W9211104
Large
Case of 6 for $365.50
Add to cart
 
17-999-927
View Documents Promotion Details
Ansell™
YE30W9211105
X-Large
Case of 6 for $365.50
Add to cart
 
17-999-928
View Documents Promotion Details
Ansell™
YE30W9211106
2X-Large
Case of 6 for $365.50
Add to cart
 
17-999-931
View Documents Promotion Details
Ansell™
YE30W9211109
5X-Large
Case of 6 for $460.50
Add to cart
 
17-999-929
View Documents Promotion Details
Ansell™
YE30W9211107
3X-Large
Case of 6 for $387.50
Add to cart
 
17-999-930
View Documents Promotion Details
Ansell™
YE30W9211108
4X-Large
Case of 6 for $418.50
Add to cart
 
Description

Description

  • Protection: Multi-layer barrier fabric effective against numerous chemicals
  • Highly visible: Bright yellow for improved worker safety
  • Anti-static: Tested according to EN1149-5 & AATCC76 (8EMSL test method)
  • Design: Elastic waist, wrists and ankles, double zip closure, double cuffs, ultrasonically welded seams, finger loops, 2 piece respirator fit hood
  • Applications: Chemicals, oil and petrochemicals, pharmaceutical, food indsutry, sewage purification installations, industrial and tank cleaning, mining
  • Sizes: Available in Small through 5X-Large
  • Certifications: EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
  • Storage: 5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B

Specifications

Specifications

No
Barrier laminate fabric
3-panel, Respirator-fit
Double, Elastic
Elastic
Liquid and spray tight chemical and biological protection
Yellow
Hooded
Double zipper
Elasticated
Welded
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit