Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-3000 Yellow Coveralls with Hood and Socks

Provide an extremely effective barrier against both chemical and biological hazards. Ansell™ Alphatec™ MICROCHEM™ 68-3000 Yellow Coveralls with Hood and Socks are durable and multi-layered, and are some of the most comfortable chemical protective materials available.

Manufacturer:  Ansell™ YE30W9212207

 View more versions of this product

Catalog No. 18-000-009


  Check Availability
Add to cart

Alert:

This product contains natural rubber latex which may cause allergic reactions.


Specifications

Description

  • Protection:  Multi-layer barrier fabric effective against numerous chemicals
  • Highly visible:  Bright yellow for improved worker safety
  • Anti-static:  Tested according to EN1149-5 & AATCC76 (8EMSL test method)
  • Design:  Elastic waist, wrists and ankles, double zip closure, double cuffs, ultrasonically welded seams, finger loops, 2 piece respirator fit hood, integrated socks with boot overflap
  • Applications:  Chemicals, oil and petrochemicals, pharmaceutical, food indsutry, sewage purification installations, industrial and tank cleaning, mining
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B

Description & Specifications

Specifications

Double zip closure
Elasticated double cuffs
Ultrasonically welded seams
3XL
No
Elasticated
Elasticated with integrated socks and boot overflaps
Barrier laminate fabric
Yellow
3 piece respirator fit hood
Attached hood
Liquid and spray tight chemical and biological protection
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit