Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-6000 Gastight Coveralls with Attached Gloves

Provide protection for emergency responders or chemical workers against dangerous and toxic chemicals in liquid or gaseous form. Ansell™ Alphatec™ MICROCHEM™ 68-6000 Gastight Coveralls with Attached Gloves are Level A suits where a self-contained breathing apparatus is worn on the inside.

Manufacturer:  Ansell™ OR60T9280105G02

 View more versions of this product

Catalog No. 18-000-023


  Check Availability
Add to cart

Specifications

Description

  • Protection:  Level A Gas tight suit tested against numerous chemicals including the ASTM F1001 test battery
  • Fabric technology:  Co-extruded multi-layer performance barrier laminate with scrim reinforcement
  • Visor technology:  Semi-rigid 3 layer PET visor providing excellent optical clarity
  • Design:  DYNAT/YKK gas-tight zipper, permanently attached Ansell Barrier gloves with sleeve over-cuffs, ultrasonically welded and taped seams
  • Applications:  Chemicals, pharmaceuticals, emergency service, sewage treatment, nuclear, petrochemicals, first response, industrial and tank cleaning, shipping
  • Sizes:  Available in Small through 3X-Large
  • Certifications:  EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN943-1 Type 1a, EN943-2 Type 1a-ET
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN943-1 Type 1a, EN943-2 Type 1a-ET

Description & Specifications

Specifications

DYNAT/YKK gas-tight zipper
Barrier laminate fabric
Attached gloves
Orange
Gas tight chemical and biological protection
Permanently attached Ansell Barrier glove with sleeve over-cuffs
Attached socks,Boot overflaps
Taped,Ultrasonically welded
X-Large
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit