Ansell™ Alphatec™ MICROCHEM™ 68-6000 Gastight Coveralls with Dual Glove Locking Cuff, internal link

Provide protection for emergency responders or chemical workers against dangerous and toxic chemicals in liquid or gaseous form. Ansell™ Alphatec™ MICROCHEM™ 68-6000 Gastight Coveralls with Dual Glove Locking Cuffs are Level A suits where a self-contained breathing apparatus is worn on the inside.

Manufacturer: Ansell™ OR60T9280204GA1

 View more versions of this product

Catalog No. 18-000-028

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Material Barrier laminate fabric
    Seam Type Ultrasonically welded and taped seams
    Color Orange
    Size Large
    Closure Type DYNAT/YKK gas-tight zipper
    Protection Class Gas tight chemical and biological protection
    Ankle Style Attached socks with boot overflaps
    Cuff Style Interchangeable locking cuff for dual glove system (Ansell Barrier and Neoprene), inner link

    • Protection:  Level A Gas tight suit tested against numerous chemicals including the ASTM F1001 test battery
    • Fabric technology:  Co-extruded multi-layer performance barrier laminate with scrim reinforcement
    • Visor technology:  Semi-rigid 3 layer PET visor providing excellent optical clarity
    • Design:  DYNAT/YKK gas-tight zipper, interchangeable locking cuff for dual glove system (internal link), ultrasonically welded and taped seams
    • Applications:  Chemicals, pharmaceuticals, emergency service, sewage treatment, nuclear, petrochemicals, first response, industrial and tank cleaning, shipping
    • Sizes:  Available in Small through 3X-Large
    • Certifications:  EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN943-1 Type 1a, EN943-2 Type 1a-ET
    • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

    Certifications

    EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN943-1 Type 1a, EN943-2 Type 1a-ET

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit