Ansell™ Alphatec™ MICROCHEM™ 68-CFR Flame Retardant Coveralls with Hood

Offer protection from particulates and chemical sprays or splashes without compromising worker protection in the event of a flashfire. Ansell™ Alphatec™ MICROCHEM™ 68-CFR Flame Retardant Coveralls are manufactured from flame retardant and antistatic fabric. They are ideal for use in industries like oil and petrochemicals, petrolium distribution and processing, and utilities.

Manufacturer: Ansell™ RD96T9211103

 View more versions of this product

Catalog No. 17-999-957

  Check Availability
Add to Cart

Alert

This product contains natural rubber latex which may cause allergic reactions. Must be worn over thermal protective garments, such as NOMEX™ or PYROVATEX™, and should never be worn next to the skin.


    Specifications

    Material CFR
    Seam Type Serged and taped seams
    Color Red
    Size Medium
    Hood Type 2 piece hood
    Latex Free No
    Neck Style Attached hood
    Closure Type Double zip closure
    Waist Style Elasticated
    Protection Class Liquid and spray tight chemical and biological protection
    Ankle Style Elasticated
    Cuff Style Elasticated wrists

    • Protection:  Flame retardant treated fabric with PVC barrier film offering wearers protection from liquid chemicals
    • Versatile:  Good for most applications where there is the need for protection from chemical spray without compromising wearer protection in the event of a flash fire
    • Optimized body fit:  Improves wearer comfort and safety
    • Highly visible:  Bright red color to improve worker safety
    • Design:  Elastic waist, wrists and ankles, double zip closure, double cuffs with knitted inner cuff, serged and taped seams, 2 piece hood
    • Applications:  Oil and petrochemicals, petroleum distribution and processing, utilities
    • Sizes:  Available in Small through 5X-Large
    • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, EN 14126 / ASTM F1671, EN 14605 Type 3, EN 14605 Type 4, EN ISO 14116 INDEX 1/0/0
    • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

    Certifications

    EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, EN 14126 / ASTM F1671, EN 14605 Type 3, EN 14605 Type 4, EN ISO 14116 INDEX 1/0/1

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit