Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-CFR Flame Retardant Coveralls with Hood

Offer protection from particulates and chemical sprays or splashes without compromising worker protection in the event of a flashfire. Ansell™ Alphatec™ MICROCHEM™ 68-CFR Flame Retardant Coveralls are manufactured from flame retardant and antistatic fabric. They are ideal for use in industries like oil and petrochemicals, petrolium distribution and processing, and utilities.

$566.75 - $1421.00

Specifications

Latex Free No
Color Red
Material CFR
Neck Style Hooded
Hood Type 2-panel
View More Specs

Alert:

This product contains natural rubber latex which may cause allergic reactions. Must be worn over thermal protective garments, such as NOMEX™ or PYROVATEX™, and should never be worn next to the skin.


Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-999-956
View Documents
Ansell™
RD96T9211102
Small
Case of 20 for $1,421.00
Add to cart
 
17-999-957
View Documents
Ansell™
RD96T9211103
Medium
Case of 20 for $1,421.00
Add to cart
 
17-999-958
View Documents
Ansell™
RD96T9211104
Large
Case of 20 for $1,421.00
Add to cart
 
17-999-959
View Documents
Ansell™
RD96T9211105
X-Large
Case of 20 for $1,421.00
Add to cart
 
17-999-960
View Documents
Ansell™
RD96T9211106
2X-Large
Case of 20 for $660.33
Add to cart
 
17-999-962
View Documents
Ansell™
RD96T9211108
4X-Large
Case of 15 for $566.75
Add to cart
 
17-999-961
View Documents
Ansell™
RD96T9211107
3X-Large
Case of 20 for $699.50
Add to cart
 
17-999-963
View Documents
Ansell™
RD96T9211109
5X-Large
Case of 15 for $1,327.00
Add to cart
 
Description

Description

  • Protection:  Flame retardant treated fabric with PVC barrier film offering wearers protection from liquid chemicals
  • Versatile:  Good for most applications where there is the need for protection from chemical spray without compromising wearer protection in the event of a flash fire
  • Optimized body fit:  Improves wearer comfort and safety
  • Highly visible:  Bright red color to improve worker safety
  • Design:  Elastic waist, wrists and ankles, double zip closure, double cuffs with knitted inner cuff, serged and taped seams, 2 piece hood
  • Applications:  Oil and petrochemicals, petroleum distribution and processing, utilities
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, EN 14126 / ASTM F1671, EN 14605 Type 3, EN 14605 Type 4, EN ISO 14116 INDEX 1/0/0
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, EN 14126 / ASTM F1671, EN 14605 Type 3, EN 14605 Type 4, EN ISO 14116 INDEX 1/0/0

Specifications

Specifications

No
CFR
2-panel
Elastic
Elastic
Liquid and spray tight chemical and biological protection
Red
Hooded
Double zipper
Elasticated
Serged, Taped
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit