Learn More
Alphaxalone, Tocris
Direct activator and potentiator of GABAA
Manufacturer:  R&D Systems 3046/50
Quantity 50mg
 View more versions of this product
Catalog No. 30-465-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
332.48
Alphaxalone
C21H32O3
(3α, 5α)-3-Hydroxypregnane-11, 20-dione
Specifications
23930-19-0
C21H32O3
(3α, 5α)-3-Hydroxypregnane-11, 20-dione
Alphaxalone
>98%
332.48
50mg
Safety and Handling
Recommended Storage : Store at Room Temperature