Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alphazurine A, Acros Organics

Click to view available options
Quantity:
10g
3486-30-4
C37H35N2NaO6S2
690.805
MFCD00044440
FTUYQIPAPWPHNC-UHFFFAOYSA-M
Acid Blue 7,C.I. 42080,Patent Blue A
62498
sodium;4-[[4-[benzyl(ethyl)amino]phenyl]-[4-[benzyl(ethyl)azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulfonate
CCN(CC1=CC=CC=C1)C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC=CC=C4)C=C3)C5=C(C=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+]
3486-30-4
C37H35N2NaO6S2
690.805
MFCD00044440
FTUYQIPAPWPHNC-UHFFFAOYSA-M
Acid Blue 7,C.I. 42080,Patent Blue A
62498
sodium;4-[[4-[benzyl(ethyl)amino]phenyl]-[4-[benzyl(ethyl)azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulfonate
CCN(CC1=CC=CC=C1)C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC=CC=C4)C=C3)C5=C(C=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+]
Beilstein 14, 856
Infrared Spectrum Authentic
Formula Weight 690.79
Physical Form Crystalline Powder
UV Spectrometry Complies
Color Purple
Quantity 10g
Chemical Name or Material Alphazurine A