Sino Biological™ ALPI Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding mature form of human ALPI (Met 1- Asp 503), w/o pro peptide, fused to polyhistidine tag at C-terminus

Manufacturer: Sino Biological™ 13225H02H5

 View more versions of this product

Catalog No. 13-225-H02H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 79.5kd
    Purity ≥90% by SDS-PAGE
    Quantity 5μg
    Source Human
    Structural Form Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name Alkaline Phosphatase Intestinal
    Gene Symbol ALPI
    Protein Tag IgG1 Fc Tag
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System Mammalian
    Product Type ALPI Recombinant Human Protein
    For Use With (Application) Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
    Format Lyophilized

    Expressed from DNA sequence encoding mature form of human ALPI (Met 1- Asp 503), w/o pro peptide, fused to polyhistidine tag at C-terminus

    • Formulation: Lyopihilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH: 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit