Learn More
ALPK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 191071AP
Catalog No. 50-173-2343
Add to cart
Description
Description
Unlike most eukaryotic kinases, alpha kinases, such as LAK, recognize phosphorylation sites in which the surrounding peptides have an alpha-helical conformation.
Specifications
Specifications
ALPK1
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96QP1, Q9CXB8
8430410J10Rik, alpha kinase 1, Alpha protein kinase 1, ALPK1, Chromosome 4 kinase, KIAA1527, LAK
Rabbit
IgG
150 μL
-20°C
Primary
310879, 71481, 80216
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ALPK1
Liquid
Alpk1
ALPK1 Fusion Protein Ag5320
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat