Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALPP Mouse anti-Human, Clone: PL8-F6, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14268

Catalog No. 89-034-350


Add to cart

Description

Description

Mouse monoclonal antibody raised against native human ALPP.

  • High specificity
  • High reactivity
Specifications

Specifications

ALPP
PL8-F6
Mouse monoclonal antibody raised against native human ALPP.
In 10 mM PBS.
Liquid
ALPP
Native purified human ALPP.
Protein A/G purification
RUO
Primary
250
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Immunohistochemistry (Frozen sections) (0.25-0.5 ug/mL) The optimal
P05187
ALP|FLJ61142|PALP|PLAP
Mouse
IgG2b, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This monoclonal antibody reacts with a membrane-bound isozyme (Regan and Nagao type) of ALPP occurring in the placenta during the 3rd trimester of gestation. It is highly specific for PLAP and shows no cross-reaction with other isozymes of alkaline phosphatase.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)