Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alprostadil, USP, 95-105%, Spectrum™

Click to view available options
Quantity:
10 mg
25 mg
100 mg
1 g
Packaging:
Amber Glass Bottle
745-65-3
C20H34O5
354.49
GMVPRGQOIOIIMI-DWKJAMRDSA-N
7-[(1R,2R,3R)-3-hydroxy-2-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-5-oxocyclopentyl]heptanoic acid
CCCCC[C@H](O)\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(O)=O
745-65-3
C20H34O5
354.49
GMVPRGQOIOIIMI-DWKJAMRDSA-N
7-[(1R,2R,3R)-3-hydroxy-2-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-5-oxocyclopentyl]heptanoic acid
CCCCC[C@H](O)\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(O)=O

P1679, 745-65-3, C20H34O5

Spectrum™ Alprostadil, USP is used to treat male sexual function problems (erectile dysfunction). All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.5%
Packaging Amber Glass Bottle
Quantity 25 mg
CAS Max % 100%
UN Number UN2811
Percent Purity 95 to 105%