Learn More
Abnova™ ALS2 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00057679T02
Catalog No. 89-933-255
Add to cart
Description
Description
Specifications
Specifications
293T
BC071576.1
pCMV-ALS2 full-length
100 μL
Human
ALS2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 135.7kDa
Transient overexpression cell lysate was tested with anti-ALS2 antibody (Mfr. No. H00057679-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot