Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALS2 Mouse anti-Mouse, Human, Bovine, Rat, Clone: N-alsin-24, MilliporeSigma™

Mouse Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen ALS2
Clone N-alsin-24
Concentration 1 mg/mL
Applications ELISA, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN2628100
View Documents
MilliporeSigma
MABN2628100UG
100 μg
Each for $387.00
Add to cart
 
MABN262825
View Documents
MilliporeSigma
MABN262825UG
25 μg
Each for $153.40
Add to cart
 
Description

Description

ALS2 Monoclonal antibody specifically detects ALS2 in Mouse, Human, Bovine, Rat samples. It is validated for ELISA, Western Blotting.
Specifications

Specifications

ALS2
1 mg/mL
Unconjugated
Mouse
Mouse, Human, Bovine, Rat
GST-tagged recombinant fragment corresponding to the first 364 amino acids from the N-terminal region of murine Alsin.
Primary
Stable for 1 year at 2-8°C from date of receipt.
N-alsin-24
ELISA, Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Monoclonal
Protein G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit