Learn More
ALS2CR3 Mouse anti-Human, Rat, DyLight 755, Clone: S390-43, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259333IR
Catalog No. NBP259333IR
Add to cart
Description
Description
ALS2CR3 Monoclonal antibody specifically detects ALS2CR3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
ALS2CR3
S390-43
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
TRAK2
Fusion protein amino acids 757-914 (C-terminus) of human TRAK2
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 100kDa. Does not cross-react with TRAK1.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
ALS2CR3GRIF1, CALS-C, GRIF-1, KIAA0549amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3, OIP98Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein, trafficking kinesin-binding protein 2, trafficking protein, kinesin binding 2
Mouse
IgG2a
100 ul
Primary
66008
Human, Rat